Frizzled-4 is a 537 amino acid protein encoded by the human gene FZD4. Frizzled-4 acts as a receptor for Wnt proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. 1 * 100 µG
Catalog # BWRLBS60963
Supplier: Bioworld Technology
Frequently Bought Together