This gene encodes a protein that is localized to mitochondria and plays a critical role in iron-sulfur cluster biogenesis. The encod ed protein assembles and transfers 4Fe-4S clusters to target apoproteins including succinate dehydrogenase and lipoic 1 * 100 µG
Catalog # BWRLBS8705
Supplier: Bioworld Technology
Frequently Bought Together