Order Entry
Sweden
ContactUsLinkComponent
SH3BGRL (SH3 domain binding glutamic acid-rich protein like), also known as SH3BGR, is a 114 amino acid protein that is ubiquitously expressed and is encoded by a gene which maps to the X chromosome of both human and mouse. 1 * 100 µG
Catalog # BWRLBS60479
Leverantör:  Bioworld Technology
SH3BGRL (SH3 domain binding glutamic acid-rich protein like), also known as SH3BGR, is a 114 amino acid protein that is ubiquitously expressed and is encoded by a gene which maps to the X chromosome of both human and mouse. 1 * 100 µG
Catalog # BWRLBS60479
Leverantör:  Bioworld Technology
Frequently Bought Together