Order Entry
Sweden
ContactUsLinkComponent
NDUFA1 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1), also known as ZNF183 or MWFE, is one of about 45 subunits comprising Complex I of the oxidative phosphorylation electron transport chain. 1 * 100 µG
Catalog # BWRLBS60474
Leverantör:  Bioworld Technology
NDUFA1 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1), also known as ZNF183 or MWFE, is one of about 45 subunits comprising Complex I of the oxidative phosphorylation electron transport chain. 1 * 100 µG
Catalog # BWRLBS60474
Leverantör:  Bioworld Technology
Frequently Bought Together