Protein disulfide isomerases (EC 5.3.4.1), such as PDIA6, are endoplasmic reticulum (ER) resident proteins that catalyze formation, reduction, and isomerization of disulfide bonds in proteins and are thought to play a role in folding of disulfide-bon 1 * 100 µG
Catalog # BWRLBS8675
Supplier: Bioworld Technology
Frequently Bought Together