This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc io n with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipe 1 * 100 µG
Catalog # BWRLBS8674
Supplier: Bioworld Technology
Frequently Bought Together