The HIRA protein shares sequence similarity with Hir1p and Hir2p, the two corepressors of histone gene transcription characterized i n the yeast, Saccharomyces cerevisiae. The structural features of the HIRA protein suggest that it may function as pa 1 * 100 µG
Catalog # BWRLBS8648
Supplier: Bioworld Technology
Frequently Bought Together