JTV1, also known as P38 or AIMP2 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 2), is a 320 amino acid nucl ear and cytoplasmic protein that is a component of the aminoacyl-tRNA synthase complex. 1 * 100 µG
Catalog # BWRLAP6009
Supplier: Bioworld Technology
Frequently Bought Together