DDX36 (DEAH box protein 36), also known as MLE-like protein 1 and RNA helicase associated with AU-rich element ARE, is a 1,008 amino acid protein that is expressed in testis and may function in sex development and spermatogenesis. 1 * 100 µG
Catalog # BWRLAP6004
Supplier: Bioworld Technology
Frequently Bought Together