Glucokinase (also designated hexokinase IV, HXKIV or GCK) plays a key role in the regulation of glucose-induced insulin secretion. G CK is expressed in pancreatic beta cells where it functions as the major glucose sensor of the body, determining the 1 * 100 µG
Catalog # BWRLBS8515
Supplier: Bioworld Technology
Frequently Bought Together