Cells undergoing apoptosis lose the asymmetry of plasma membrane phospholipids, and phosphatidylserine is exposed on the outer surfa ce of the membrane. The phosphatidylserine receptor (PSR) specifically recognizes phosphatidylserine . 1 * 100 µG
Catalog # BWRLBS8501
Supplier: Bioworld Technology
Frequently Bought Together