Olfactory receptors interact with odorant molecules in thenose, to initiate a neuronal response that triggers the perceptionof a sme ll. The olfactory receptor proteins are members of a largefamily of G-protein-coupled receptors (GPCR) arising from s 1 * 100 µG
Katalog # BWRLBS61502
Supplier: Bioworld Technology
Frequently Bought Together