Order Entry
Sweden
ContactUsLinkComponent
Class E vacuolar protein sorting (vps) proteins are necessary for appropriate sorting of receptors in the yeast endocytic pathway. T he yeast Vps4p is a member of the AAA protein family (ATPases associated with diverse cellular activities). 1 * 100 µG
Catalog # BWRLBS61477
Leverantör:  Bioworld Technology
Class E vacuolar protein sorting (vps) proteins are necessary for appropriate sorting of receptors in the yeast endocytic pathway. T he yeast Vps4p is a member of the AAA protein family (ATPases associated with diverse cellular activities). 1 * 100 µG
Catalog # BWRLBS61477
Leverantör:  Bioworld Technology
Frequently Bought Together