c-Jun N-terminal kinases (JNKs) phosphorylate and augment transcriptional activity of c-Jun. JNKs originate from three genes that yi eld 10 isoforms through alternative mRNA splicing. 1 * 100 µG
Catalog # BWRLMB0184
Supplier: Bioworld Technology
Frequently Bought Together