Order Entry
Sweden
ContactUsLinkComponent
THE SLURRY GLASS CONCENTRIC NEBULIZER IS IDEAL FOR SPECIALIZED APPLICATIONS REQUIRING THE ASPIRATION OF SLURRIES OR SUSPENSIONS. ITS CAPILLARY BORE IS 50% LARGER THAN THAT OF STANDARD CONCENTRIC NEBULIZERS AND THIS REDUCES THE POSSIBILITY OF PARTICL 1 * 1 items
Catalog # PERCN8121059
Supplier:  PerkinElmer
THE SLURRY GLASS CONCENTRIC NEBULIZER IS IDEAL FOR SPECIALIZED APPLICATIONS REQUIRING THE ASPIRATION OF SLURRIES OR SUSPENSIONS. ITS CAPILLARY BORE IS 50% LARGER THAN THAT OF STANDARD CONCENTRIC NEBULIZERS AND THIS REDUCES THE POSSIBILITY OF PARTICL 1 * 1 items
Catalog # PERCN8121059
Supplier:  PerkinElmer
Frequently Bought Together