0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HYDROMETER F.MILK OF LIME 0-20% IN 0.5% 1 * 1 items
Catalog # AMARSF-H000345
Supplier:
Amarell
Expand image
HYDROMETER F.MILK OF LIME 0-20% IN 0.5% 1 * 1 items
Catalog # AMARSF-H000345
Supplier:
Amarell