Order Entry
Sweden
Orders LinkContactUsLinkComponent
Spore ampoules

Spore ampoules

Geobacillus stearothermophilus cell line 7953: spore ampoule104 (24 month shelf life) / sterilisation process: steam liquid cycles, 58 mm, 1 ml
Catalog # NAMSSA1-50-04
Supplier:   CROSSTEX
Spore ampoules
Catalog # NAMSSA1-50-04
Supplier:  CROSSTEX
Spore ampoules

Geobacillus stearothermophilus cell line 7953: spore ampoule104 (24 month shelf life) / sterilisation process: steam liquid cycles, 58 mm, 1 ml

Price
3 675,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

These spore ampoules are ideal for use in validation and routine monitoring of liquid steam sterilization cycles. Spore ampoules are manufactured using hermetically sealed Type I borosilicate glass containing an exclusively formulated Tryptic
Soy Broth (TSB) modified with pH indicator, and Geobacillus stearothermophilus spores. No activation is required; simply expose Spore Ampoules to sterilization process and incubate.

  • Reduced incubation time of 48 hours
  • Clear visible results as growth is evident by turbidity and/or a color shift of the media from purple to yellow
  • Saves time by eliminating aseptic culturing required with traditional spore strips
  • Unique packaging insert keeps ampoules in upright position and allows for quick reference to the number of units remaining

Delivery information: Each shipment is accompanied by a certificate of analysis and instructions for use

Caution: Storage under refrigerated conditions (2 to 8 °C) is required. Do not freeze spore ampoules.

Documents

Specifications
  • Volume:
    1 ml
  • Description:
    Geobacillus stearothermophilus cell line 7953: spore ampoule104 (24 month shelf life) / sterilisation process: steam liquid cycles
  • Length:
    58 mm
  • Seq:
    05