Order Entry
Sweden
ContactUsLinkComponent
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-mouse IgG (H+L) with near-IR fluorescent dye CF™ 750ST for (d)-STORM super-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed again e 1 * 500 µl
Catalog # 20825-500UL
Supplier:  Biotium
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-mouse IgG (H+L) with near-IR fluorescent dye CF™ 750ST for (d)-STORM super-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed again e 1 * 500 µl
Catalog # 20825-500UL
Supplier:  Biotium
Frequently Bought Together