LigaTrap™ resins bind an exceptionally broad range of recombinant, monoclonal, and human/animal source polyclonal immunoglobulins. L igaTrap™ resins provide distinct advantages to traditional bind and elute purifications resins including mild elution 1 * 1 KIT
Catalog # LIGALT-095KIT
Supplier: LIGATRAP TECHNOLOGIES