Order Entry
Sweden
ContactUsLinkComponent
Reduction column, funnel on the top d=50 mm, h=45 mm, column tube o.d. 26 mm, wall 2,0 mm, PTFE stopcock NS 12,5-2,5 mm, draining tube o.d. 8 mm, L=100 mm, with pointed tip, length from stopcock to the upper end approx. 370 mm, acc to. drawing, made 1 * 1 items
  LENZ81003386
Reduction column, funnel on the top d=50 mm, h=45 mm, column tube o.d. 26 mm, wall 2,0 mm, PTFE stopcock NS 12,5-2,5 mm, draining tube o.d. 8 mm, L=100 mm, with pointed tip, length from stopcock to the upper end approx. 370 mm, acc to. drawing, made 1 * 1 items
  LENZ81003386