(±)-1,2-Propanediol, liquid
Product Details & Documents
Propylene glycol is a clear, colorless and hygroscopic liquid. Propylene glycol contains an asymmetrical carbon atom, so it exists in two stereoisomers.
The freezing point of water is depressed when mixed with propylene glycol due to increased opportunity for hydrogen bonding.
Propylene glycol is a solvent in many pharmaceuticals, including oral, injectable and topical formulations, such as for diazepam and lorazepam that are insoluble in water, use propylene glycol as a solvent in their clinical, injectable forms. It is also used as automotive antifreeze.
- CAS-nummer:57-55-6
- Formel:CH₃CH(OH)CH₂OH
- MDL-nummer:MFCD00064272
- Molecular Weight:76.1 g/mol
- Kokande Pt:187.9 °C (1013 hPa) (lit.)
- Smält Pt:–60 °C (lit.)
- Densitet:1.04 g/cm³ (20 °C) (lit.)
- Förvaringstemperatur:Ambient
Specifikationer
Specifikationstestresultat
- Identity Test:Passes
- FTIR:Conforms to Standard
Specifikationer
- Pk:1 L
Specifikationer
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



