Order Entry
Sweden
ContactUsLinkComponent
Microscope cover glasses, 73x109 mm, borosilicate glass D 263, thickness No. 1.5 (0.16-0.19 mm), paper-interleaved, 50 pcs./cardboard box, SUPERIOR 1 * 1.000 items
Catalog # MARI01029990932
Supplier:  MARIENFELD
Microscope cover glasses, 73x109 mm, borosilicate glass D 263, thickness No. 1.5 (0.16-0.19 mm), paper-interleaved, 50 pcs./cardboard box, SUPERIOR 1 * 1.000 items
Catalog # MARI01029990932
Supplier:  MARIENFELD
Frequently Bought Together