Order Entry
Sweden
Orders LinkContactUsLinkComponent
VWR®, Microtube Storage Boxes
100, Orange, 141×151×47 mm
Katalog # 525-0931
Leverantör:   VWR Collection
VWR®, Microtube Storage Boxes
Katalog # 525-0931
Leverantör:  VWR Collection
VWR®, Microtube Storage Boxes
VWR®, Microtube Storage Boxes
100, Orange, 141×151×47 mm
Price
1 075,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 5 Items
Product Details & Documents
PP.
  • Highly transparent, with bold identifying colours and a distinctive wave design
  • Uniform wells with location ID; capacity indicated on the outside of the box
  • Black light-tight version available for light-sensitive samples
  • Frosted labelling patch
  • Alphanumeric identification on the bottom
Compact footprint for microtube and cryogenic vial storage. Store 1,5/2,0 ml microtubes or cryogenic vials. Complete with an easy open, friction-fit lid. Will fit into most standard freezer racking.
Delivery information: Assorted pack contains one of each colour blue, green, pink, yellow and orange.
Specifikationer
  • Compartments:
    100
  • W×D×H ext.:
    141×151×47 mm
  • Colour:
    Orange
  • Seq:
    16