Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Diethylene glycol

Poly drum, 20 L
Catalog # SPCMD1027-20LT
Irritant
Supplier:   Spectrum Chemical
Diethylene glycol
Catalog # SPCMD1027-20LT
Supplier:  Spectrum Chemical
CAS Number:  111-46-6
Diethylene glycol
Diethylene glycol

Poly drum, 20 L

CAS Number:  111-46-6
Price
Inquire For Price
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol
  • CAS Number:111-46-6
  • Formula:C₄H₁₀O₃
  • MDL Number:MFCD00002882
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,03146
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:–10 °C
  • Density:1.118 g/cm³ (20 °C)
  • Flash Pt:123 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-872-2
Specifications

Specification Test Results

  • Assay:
    97% Min.

Specifications

  • Pack type:
    Poly drum
  • Pk:
    20 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.