Order Entry
Sweden
ContactUsLinkComponent
MTC306 Silver combined electrode, 1 m cable, for titration use with TITRALAB AT1000 Series 1 * 1 items
Catalog # HACHMTC30601
Supplier:  Hach
MTC306 Silver combined electrode, 1 m cable, for titration use with TITRALAB AT1000 Series 1 * 1 items
Catalog # HACHMTC30601
Supplier:  Hach