Order Entry
Sweden
Orders LinkContactUsLinkComponent
Water hardness testing solutions
Water hardness testing solutions
Hardness 3 solution (EDTA), in a dropping bottle, EDTA, 500 mL
Catalog # HACH42649
Supplier:   Hach
Water hardness testing solutions
Catalog # HACH42649
Supplier:  Hach
CAS Number:  
Water hardness testing solutions
Water hardness testing solutions
Hardness 3 solution (EDTA), in a dropping bottle, EDTA, 500 mL
CAS Number:  
Price
605,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications
Specifications
  • Chemical method:
    EDTA
  • Pk:
    500 mL
  • Description:
    Hardness 3 solution (EDTA), in a dropping bottle
Specifications