Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
METHANOIC ACID 85 % TECHNICAL 1 * 1.200 kg
Catalog # SWEDMY843802
Leverantör:
SWED HANDLING
Expand image
METHANOIC ACID 85 % TECHNICAL 1 * 1.200 kg
Catalog # SWEDMY843802
Leverantör:
SWED HANDLING