0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Biacore Series S compatible. Coating: 200 nm biotin derivatized linear polycarboxylate hydrogel, medium charge density. 1 * 5 items
Catalog # MSPPSCBSBHC200M-5
Supplier:
MARKET SOURCE PART PROCESS
Expand image
Biacore Series S compatible. Coating: 200 nm biotin derivatized linear polycarboxylate hydrogel, medium charge density. 1 * 5 items
Catalog # MSPPSCBSBHC200M-5
Supplier:
MARKET SOURCE PART PROCESS