Order Entry
Sweden
Orders LinkContactUsLinkComponent
Ribbons for label printer BBP™11

Ribbons for label printer BBP™11

Thermal transfer printer ribbon, 4900 Series, width 110 mm, BBP™11, 70 m, Black
Catalog # BRDY804795
Supplier:   Brady
Ribbons for label printer BBP™11
Catalog # BRDY804795
Supplier:  Brady
Ribbons for label printer BBP™11
Ribbons for label printer BBP™11

Thermal transfer printer ribbon, 4900 Series, width 110 mm, BBP™11, 70 m, Black

Price
570,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Colour:
    Black
  • Description:
    Thermal transfer printer ribbon, 4900 Series, width 110 mm
  • Length:
    70 m
  • Seq:
    20
  • For:
    BBP™11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.