Order Entry
Sweden
ContactUsLinkComponent
Sipper for Libra S50/60/70/80. Requires Tubing kit(80-7100-37 or 80-7100-38) & suitable flowcell (80-7100-40 recommended) 1 * 1 items
Catalog # WAPR80-7100-36
Supplier:  Biochrom
Sipper for Libra S50/60/70/80. Requires Tubing kit(80-7100-37 or 80-7100-38) & suitable flowcell (80-7100-40 recommended) 1 * 1 items
Catalog # WAPR80-7100-36
Supplier:  Biochrom