Order Entry
Sweden
Orders LinkContactUsLinkComponent
Pyridine 99+% for analysis

Pyridine 99+% for analysis

Glass bottle, 1 L
Catalog # ACRO290860010
Irritant
Flammable
Pyridine 99+% for analysis
Catalog # ACRO290860010
CAS Number:  110-86-1
Pyridine 99+% for analysis
Pyridine 99+% for analysis

Glass bottle, 1 L

CAS Number:  110-86-1
Price
2 700,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Boiling Pt:115…116 °C (760 mmHg)
  • Melting Pt:–42 °C
  • Density:0.978 g/cm³
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • UN:1282
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Non-volatile matter:
    ≤0.002 %
  • Ammonia (NH3):
    ≤0.002 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications