Order Entry
Sweden
Orders LinkContactUsLinkComponent
Ribbons for label printer BBP™11

Ribbons for label printer BBP™11

Thermal transfer printer ribbon, 7953 series, width 65 mm, BBP™11, 70 m, Black
Catalog # BRDY804450
Supplier:   Brady
Ribbons for label printer BBP™11
Catalog # BRDY804450
Supplier:  Brady
Ribbons for label printer BBP™11
Ribbons for label printer BBP™11

Thermal transfer printer ribbon, 7953 series, width 65 mm, BBP™11, 70 m, Black

Price
260,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Colour:
    Black
  • Description:
    Thermal transfer printer ribbon, 7953 series, width 65 mm
  • Length:
    70 m
  • Seq:
    07
  • For:
    BBP™11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.