Order Entry
Sweden
ContactUsLinkComponent
lead-through - preparation for 2x ermeto-fitting, enclosed, for customer assembly, sheet steel, plastic laminated plain, ral 7035 li ght grey for medium and tall cabinets with width 600/1200 mm for model(s): g30, g90 1 * 1 items
Catalog # MSGA31892
Supplier:  asecos
lead-through - preparation for 2x ermeto-fitting, enclosed, for customer assembly, sheet steel, plastic laminated plain, ral 7035 li ght grey for medium and tall cabinets with width 600/1200 mm for model(s): g30, g90 1 * 1 items
Catalog # MSGA31892
Supplier:  asecos
Frequently Bought Together