Hydrochloric Acid 0.5 mol/l (0.5N) volumetric solution 1 * 5 L
Catalog # APPC181022.1214
Supplier: PanReac AppliChem
Hydrochloric Acid 0.5 mol/l (0.5N) volumetric solution 1 * 5 L
Catalog # APPC181022.1214
Supplier: PanReac AppliChem
Frequently Bought Together
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals