Order Entry
Sweden
ContactUsLinkComponent
G-Biosciences Bolton-Hunter Reagent conjugates tyrosine-like groups to end-terminal -amino groups or -amino groups of lysine to i ncrease the number of tyrosyl groups that can be iodinated by iodine-125 labeling procedures.Radioactive iodine (125I) 1 * 1 g
Catalog # GENOBC84
Supplier:  G-Biosciences
G-Biosciences Bolton-Hunter Reagent conjugates tyrosine-like groups to end-terminal -amino groups or -amino groups of lysine to i ncrease the number of tyrosyl groups that can be iodinated by iodine-125 labeling procedures.Radioactive iodine (125I) 1 * 1 g
Catalog # GENOBC84
Supplier:  G-Biosciences
Frequently Bought Together