Order Entry
Sweden
ContactUsLinkComponent
Agarose IV is a linear polymer with a very high molecular weight, giving gel structures unlike those of traditional agaroses. This characteristic produces a strong intercatenary interaction, yeilding a gel with very high gel strength and higher exclu 1 * 500 g
Catalog # GENORC-036
Supplier:  G-Biosciences
Agarose IV is a linear polymer with a very high molecular weight, giving gel structures unlike those of traditional agaroses. This characteristic produces a strong intercatenary interaction, yeilding a gel with very high gel strength and higher exclu 1 * 500 g
Catalog # GENORC-036
Supplier:  G-Biosciences
Frequently Bought Together