Order Entry
Sweden
ContactUsLinkComponent
G-Biosciences’ Amine Magnetic Beads are 3µm, uniform magnetic beads with an amine (-NH2) surface functional group. The magnetic bead s consist of a single-crystal Fe3O4 sphere core and dextran coating layer. Through chemical modification of dextran, 1 * 5 mL
Catalog # GENO786-907
Supplier:  G-Biosciences
G-Biosciences’ Amine Magnetic Beads are 3µm, uniform magnetic beads with an amine (-NH2) surface functional group. The magnetic bead s consist of a single-crystal Fe3O4 sphere core and dextran coating layer. Through chemical modification of dextran, 1 * 5 mL
Catalog # GENO786-907
Supplier:  G-Biosciences