G-Biosciences’ Carboxyl Magnetic Beads are 3µm, uniform magnetic beads with surface functional group -COOH. The magnetic bead consis ts of a single-crystal Fe3O4 sphere core and dextran coating layer. Through chemical modification of dextran, the car 1 * 1 mL
Catalog # GENO786-908
Supplier: G-Biosciences
Frequently Bought Together