Sodium bicarbonate, Corning®
Sodium bicarbonate, Liquid 7.5% (w/v) Solution, 6×100 ml, 6
Supplier: Corning
Jump to:
Product Details & Documents
Sodium bicarbonate is used as part of a buffering system commonly used to maintain physiological pH 7,2 to 7,4 in a culture environment
Bicarbonate is a weak buffer with that makes maintaining a pH range of 7,2 to 7,6 somewhat difficult, but it is non-toxic, and is a nutrient source.
Documents
Specifications
Specifications
- Pk:6
- Description:Sodium bicarbonate, Liquid 7.5% (w/v) Solution
- Pk Info:6×100 ml
- Cat. No.:25-035-CI
- Storage temperature:Ambient
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



