Order Entry
Sweden
ContactUsLinkComponent
qPCR detection of Ureaplasma urealyticum. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-U.UREALYT
Supplier:  PRIMERDESIGN
qPCR detection of Ureaplasma urealyticum. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-U.UREALYT
Supplier:  PRIMERDESIGN
Frequently Bought Together