Order Entry
Sweden
ContactUsLinkComponent
qPCR detection of Prevotella intermedia. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-P.INTERME
Supplier:  PRIMERDESIGN
qPCR detection of Prevotella intermedia. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-P.INTERME
Supplier:  PRIMERDESIGN
Frequently Bought Together