Order Entry
Sweden
ContactUsLinkComponent
qPCR detection of orf virus (Contagious pustular dermatitis). Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-ORFV-
Supplier:  PRIMERDESIGN
qPCR detection of orf virus (Contagious pustular dermatitis). Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-ORFV-
Supplier:  PRIMERDESIGN
Frequently Bought Together