Order Entry
Sweden
ContactUsLinkComponent
qPCR detection of Mumps virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-MUMPS
Supplier:  PRIMERDESIGN
qPCR detection of Mumps virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-MUMPS
Supplier:  PRIMERDESIGN