Order Entry
Sweden
ContactUsLinkComponent
Versatile, top spec Reverse Transcription Kit supplied as separate components. Suitable for 2 nanograms - 2 micrograms of RNA templa te. 150rxn 1 * 150 Tests
  NANOSCRIPT150
Versatile, top spec Reverse Transcription Kit supplied as separate components. Suitable for 2 nanograms - 2 micrograms of RNA templa te. 150rxn 1 * 150 Tests
  NANOSCRIPT150