Order Entry
Sweden
ContactUsLinkComponent
qPCR detection of tellurite resistant Escherichia coli. Contains: Primer and probe, RT target specific primers (RNA viruses only), S tandard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-E.COLI-TE
Supplier:  PRIMERDESIGN
qPCR detection of tellurite resistant Escherichia coli. Contains: Primer and probe, RT target specific primers (RNA viruses only), S tandard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-E.COLI-TE
Supplier:  PRIMERDESIGN