Fusidate sodium
5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 751-94-0 Molecular Formula: C31H47NaO6 Formula Weight: 538.69 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: White to off-white Solubility: Soluble in water MDL No.: MFCD09054714
Applications: Targets gram positive bacteria namely those which cause skin infections
Synonyms: Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate
- CAS Number:751-94-0
- Formula:C₃₁H₄₇NaO₆
- MDL Number:MFCD09054714
- Molecular Weight:538.701 g/mol
- Merck Index:14,4317
- EINECS:212-030-3
Specifications
Specifications
- Pk:5 g



