Order Entry
Sweden
ContactUsLinkComponent
GRO Alpha Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 73 amino acids and having a molecular mass of 7811 Dalton. The GRO-alpha is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80128-5
Supplier:  Biorbyt
GRO Alpha Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 73 amino acids and having a molecular mass of 7811 Dalton. The GRO-alpha is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80128-5
Supplier:  Biorbyt
Related Information