Order Entry
Sweden
ContactUsLinkComponent
Sandwich Elisa kit for Quantitative Detection for Rat IGF-2, Insulin-like growth factor II is also known as somatomedin A. IGF-2 is a member of the insulin family of polypeptide growth factors that is involved in development and growth.1 It 1 * 96 Tests
Catalog # BIRBORB76039-96
Supplier:  Biorbyt
Sandwich Elisa kit for Quantitative Detection for Rat IGF-2, Insulin-like growth factor II is also known as somatomedin A. IGF-2 is a member of the insulin family of polypeptide growth factors that is involved in development and growth.1 It 1 * 96 Tests
Catalog # BIRBORB76039-96
Supplier:  Biorbyt
Related Information