Order Entry
Sweden
ContactUsLinkComponent
Sandwich Elisa kit for Quantitative Detection for Human TFPI, TFPITissue factor pathway inhibitor, also known as EPI, LACI or TFPI1, is a single-chain polypeptide which can reversibly inhibit Factor Xa Xa and Thrombin Factor IIa. The gene f 1 * 96 Tests
Catalog # BIRBORB96794-96
Supplier:  Biorbyt
Sandwich Elisa kit for Quantitative Detection for Human TFPI, TFPITissue factor pathway inhibitor, also known as EPI, LACI or TFPI1, is a single-chain polypeptide which can reversibly inhibit Factor Xa Xa and Thrombin Factor IIa. The gene f 1 * 96 Tests
Catalog # BIRBORB96794-96
Supplier:  Biorbyt
Related Information