0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HIV-1 p24 recombinant is 24 kDa non-glycosylated polypeptide chain. 1 * 2 µG
Catalog # BIRBORB81699-2
Supplier:
Biorbyt
Expand image
HIV-1 p24 recombinant is 24 kDa non-glycosylated polypeptide chain. 1 * 2 µG
Catalog # BIRBORB81699-2
Supplier:
Biorbyt
Related Information
Documentation
Datasheet
orb81699
Documentation